Distilling heads
Supplier: CHEMGLASS
CG-1239-02
CGLACG-1239-02EA 686 SGD
CGLACG-1239-02
Distilling heads
Distillation Components Distillation Heads
Designed for use with small quantities where hold-up between the flask and receiver must be kept to a minimum.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The thermometer bulb and stem serve as column packing. Upper hose connections are for cooling lines while the lower connection is for vacuum. Available with Vigreux indentations to improve efficiency. Also available vacuum-jacketed to further improve efficiency. The NS 14/20 size accepts a 1" immersion NS 10/18 thermometer.
Order Now
SPECIFICATIONS
Environmentally Preferable