Fritted support assemblies
Supplier: CHEMGLASS
CG-1420-01
CGLACG-1420-01EA 1130 SGD
CGLACG-1420-01
Fritted support assemblies
Filters Membrane Filter Units
Glassware system for vacuum filtration using 47 mm membranes. Assembly is designed for use with vacuum manifolds or optional plastic coated filtering flask. Support base has a sealed-in coarse porosity fritted disc.
Delivery information: Complete assembly includes a 300 ml graduated funnel, aluminium clamp, support base and silicone stopper.
Order Now
SPECIFICATIONS
Environmentally Preferable