Flasks, round bottom, glass, clear, standard ground joint, one neck
Supplier: CHEMGLASS
CG-1506-17
CGLACG-1506-17EA 136 SGD
CGLACG-1506-17
Flasks, round bottom, glass, clear, standard ground joint, one neck
Flasks Round Bottom Flasks
Heavy walled, single short neck round bottom flasks with standard taper outer joints.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes