Evaporating flasks
Supplier: CHEMGLASS
CG-1512-01 CG-1512-03 CG-1512-05 CG-1512-51 CG-1512-53 CG-1512-55
CGLACG-1512-01EA 125 SGD
CGLACG-1512-01 CGLACG-1512-03 CGLACG-1512-05 CGLACG-1512-51 CGLACG-1512-53 CGLACG-1512-55
Evaporating flasks
Flasks Rotary Evaporator Flasks
Made of borosilicate glass, coated.
  • Flasks with 24/40 joints have a square reniforcing bead
  • Plastic coated to prevent injury from broken glass, and also prevents spillage of hazardous materials
  • Very good chemical resistance
These single neck recovery flask can be used as an evaporating flask in rotary evaporators.
The flask have a heavy wall for vaccum applications and are balance to prevent run-out during rotation.