Septum stoppers, rubber, Suba-Seal®
Supplier: CHEMGLASS
CG-3024-09
CGLACG-3024-09EA 664 SGD
CGLACG-3024-09
Septum stoppers, rubber, Suba-Seal®
Stoppers Rubber Stoppers
Suba-Seal® septum stoppers are flanged stoppers with a hollow plug and sleeve extension that folds down over the neck of the vessel.
  • Made from natural red rubber
In addition to the pull over sleeve, Suba-Seal® stoppers have a series of annular serrations on the plug portion of the septa. Each of these serrations depress along the inner wall of the glass vessel to form an extremely effective air-tight seal.