O-Rings for flat flanges
Supplier: CHEMGLASS
CG-305-210
CGLACG-305-210EA 48.5 SGD
CGLACG-305-210
O-Rings for flat flanges
Seals Sealing Rings
Viton O-ring is stocked in brown 75 durometer. Ideal for acids, aliphatic hydrocarbons, oxidising agents and all petroleum products.
  • Temperature range: –30 to 200° C
  • Low compression set and high temperature resistance
  • Especially good for vacuum service and low gas permeability