Laboratory jacks, Economy
Supplier: CHEMGLASS
CG-3052-01
CGLACG-3052-01EA 479 SGD
CGLACG-3052-01
Laboratory jacks, Economy
Laboratory Jacks
These multipurpose, lightweight jacks are used in supporting hot plates, burners, oil baths, and glassware along with many other items in the lab.
  • Made of chemically resistant anodised aluminum
  • Scissor jack principle for quick and accurate height adjustment with a single turn of the control knob