Total immersion thermometers
Supplier: CHEMGLASS
CG-3508-N-02
CGLACG-3508-N-02EA 310 SGD
CGLACG-3508-N-02
Total immersion thermometers
Thermometers Analogue Thermometers
Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury.
  • Thermometer has a white back, a 6 to 7 mm outer diameter, and a standard taper 10/18 inner ground joint
  • With 1° subdivisions
Immersion is measured from the bottom of the joint to the bottom of non-mercury bulb.