Filter Funnel, Disposable
Supplier: CHEMGLASS
OP-6602-14
CGLAOP-6602-14EA 1060 SGD
CGLAOP-6602-14
Filter Funnel, Disposable
Funnels Filtration Funnels Filter Funnels
This unique disposable filter funnel system for solid-liquid separation can be used in chemistry and biological laboratories. Polypropylene filter funnel with polyethylene fritted disc.
  • Good temperature resistance
  • Good chemical resistance