Power supplies, programmable, nanoPAC MINI
Supplier: CLEAVER SCIENTIFIC
NANOPAC-300
CLEVNANOPAC-300EA 0 SGD
CLEVNANOPAC-300
Power supplies, programmable, nanoPAC MINI
Power Supplies
The programmable power supply, nanoPAC mini, is capable of running all horizontal multiSUB systems and vertical omniPAGE mini gels, either on a continuous run or timed setting up to 999 minutes. Its user-friendly interface is easily adjustable in 1 V and 1 mA increments, making it perfect for separations where precise settings are required, while its ultra compact size and two pairs of parallel power terminals, which can run two electrophoresis units simultaneously, save time and bench space.
  • 3-digit LED
  • Wipe clean polycarbonate housing
  • Dual voltage compatibility
  • Equipped with timer,
More Product Information