Cleanroom wipes, Polywipe Light
Supplier: CONTEC
7070-99
CONT7070-99EA 85.2 SGD
CONT7070-99
Cleanroom wipes, Polywipe Light
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Dry wipes designed for durability and ease of use that are ideal for cage and equipment wipe down or downtime 'super cleaning'.
  • Polyester interlock knit
  • Knife-cut edge
  • Product is double bagged to ensure product integrity
  • Lot numbers provide traceability
  • Economical bulk packaging
The Polywipe Light wipe is an unlaundered standardweight polyester interlock knit wipe with knife-cut edges that is to be used in less than critical applications (in environments of ISO Class 6 and higher). The wipes have the same basic functional characteristics as launderedMore Product Information