Aerospace cleaning wipes, Sontara® AC™
Supplier: CONTEC
AC12165
CONTAC12165EA 232 SGD
CONTAC12165
Aerospace cleaning wipes, Sontara® AC™
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
Sontara AC is a white non woven creped wipe made with 55% special cellulose and 45% polyester.
  • Extremely low lint: Leave surfaces clean
  • High strength, good absorbency and versatility
  • Contain no binders, solvent resistant
  • Durable: Wipes maintain their strength, even when wet
Ideal for a wide range of critical cleaning tasks essential to industries such as biotech, pharmaceutical and electronics, they are very effective in applications such as surface preparation for coatings, machining operations, and general maintenance. Also suitable for pre-painting tasks.