Wipes for general applications, Sontara® AC™
Supplier: CONTEC
AC9165
CONTAC9165EA 564 SGD
CONTAC9165
Wipes for general applications, Sontara® AC™
Wipes Reusable Cleaning Cloths Cloths for General Applications
Launderable non-woven wipes made from DuPont™ Sontara AC fabric. Designed for aircraft manufacturing, these wipes offer benefits to numerous manufacturing applications particularly cleaning and polishing bright metallic surfaces. Interleaved polyester/cellulose blend with creped texture and a basis weight of 74,6 g/m². No metal particles.
  • Highly absorbent, low-linting for snag resistance
  • Able to withstand many solvents without falling apart
  • Manufactured with no contaminating binders or glue
  • Wipes come in pop-up boxes for easy dispensing
Order Now
SPECIFICATIONS
Environmentally Preferable