Cleanroom wipes, sterile, SterileSorb™
Supplier: CONTEC
C2-1212IR
CONTC2-1212IREA 955 SGD
CONTC2-1212IR
Cleanroom wipes, sterile, SterileSorb™
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Non woven, hydroentangled polyester/cellulose blend. Ideal for wipe down of products prior to their introduction to the cleanroom.
  • Designed for non abrasive applications such as wiping and absorbing spills in the cleanroom
  • Absorbent, durable, and low in extractables
  • Wipes have a SAL (Sterility Assurance Level) of 10⁻⁶
Certifications: AAMI/ISO 11137 Method 1 Guidelines
Caution: Data accurate at time of publishing, latest test results and test methods can be found on supplier’s website.