Disinfectant wipes, Sporicidin®
Supplier: CONTEC
CAN-18012F
CONTCAN-18012FEA 374 SGD
CONTCAN-18012F
Disinfectant wipes, Sporicidin®
Wipes Disposable Wipes and Towels Disinfectant Wipes
Sporicidin® disinfectant wipes clean, disinfect, and deodorise. These OSHA compliant disinfectant wipes are ideal for cleaning/restoration applications, such as equipment disinfection. Recommended for use in hospitals, and medical, dental, and veterinary clinics.
  • Strong fabric for multiple surface cleaning and disinfection
  • 6 month residual activity against odor causing bacteria
  • Alcohol-free and bleach-free formula
  • Mild alkaline pH (8,5)
  • Non corrosive, non abrasive and non staining: Compatible with plastics, glass, and metals
Sporicidin is based onMore Product Information