Cleanroom wipes, non pyrogenic, sterile, Amplitude™ Kappa™
Supplier: CONTEC
NWPZ0001
CONTNWPZ0001EA 1310 SGD
CONTNWPZ0001
Cleanroom wipes, non pyrogenic, sterile, Amplitude™ Kappa™
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
These non pyrogenic LE wipes are the first sterile non woven wiping products designed for the pharmaceutical, medical device and biotech industries. Made with Sontara® lyocell/polyester blend fabric, these low endotoxin wipes have excellent sorbency and are packaged, gamma sterilised and validated sterile to a 10⁻⁶
Sterility Assurance Level.
  • Ideal for product wipe down and cleaning the exterior of equipment and facilities cleaning
  • Ideal for use in aseptic processing areas
  • Endotoxin level guaranteed less than 20 EU/wipe
  • Linear tear packaging allows for easy
More Product Information