Cleanroom wipes, pre-saturated, sterile, PROSAT® Sterile™
Supplier: CONTEC
PS-PR-7030EB
CONTPS-PR-7030EBEA 1310 SGD
CONTPS-PR-7030EB
Cleanroom wipes, pre-saturated, sterile, PROSAT® Sterile™
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These non woven wipes are designed for pharmaceutical, biotech, and medical device applications. Choose between economical hydroentangled polyester/cellulose wipes or meltblown polypropylene wipes for improved durability.
  • Designed for sterile environments
  • Presaturated wipes are convenient and safer to use than liquid IPA
  • Compact pouch easily fits into isolators, bio-safety cabinets and glove boxes
Ideal for cleanup following BD RODAC™ plate sampling, as well as for wiping isolators, cabinets, hoods, and other cleanroom equipment.
Suitable for useMore Product Information