Cleanroom wipes, pre-saturated, SATWipes® C3
Supplier: CONTEC
SAT-C3-100
CONTSAT-C3-100EA 2280 SGD
CONTSAT-C3-100
Cleanroom wipes, pre-saturated, SATWipes® C3
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
The SATWipes are made of pure non woven polyester that is hydraulically entangled creating a soft, durable and clean wipe. The saturation of either alcohol/deionised water or 100% alcohol provides a safe and mild cleaning solution.
  • Convenient and re-usable pop-up dispenser
  • Consistent solvent application and use: Up to 35% reduction in solvent use
  • Reduces solvent handling: Increased safety
Excellent for less critical applications, cleaning of process tools and other equipment and surface preparation.
Suitable for use in Class 1000, ISO 6, M4.5More Product Information