CLASSIQSwabs™ Sterile Dry Fiber Wrapped Swabs
Supplier: Copan
164KS01 155C 160C 167KS01 167KS02 168C 170KS01 184CS01 185CS01 159C 155CS01 167KS100 175KS01 162C 167CS01
COPA164KS01EA 0 SGD
COPA164KS01 COPA155C COPA160C COPA167KS01 COPA167KS02 COPA168C COPA170KS01 COPA184CS01 COPA185CS01 COPA159C COPA155CS01 COPA167KS100 COPA175KS01 COPA162C COPA167CS01
CLASSIQSwabs™ Sterile Dry Fiber Wrapped Swabs
Swabs Swabs for Sampling
CLASSIQSwabs™ are traditional and sterile ready-for-use devices intended for the collection of clinical samples.  The association of Copan CLASSIQSwabs™ with the wide range of Copan Transport Media enables an efficient sample collection and preservation compatible with many downstream assays.
These soft fiber swabs are available in tubes or peel pouches, Regular or Minitip shapes, and various materials such as cotton, rayon, and polyester. All these materials are inert and non-toxic to microorganisms and patients. CLASSIQSwabs™ are traditional and sterile ready-for-use devices intended
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable