Lab Consumables
Supplier: Copan
310C25T 311C25T 312C25T 313C25T 316C05T 317C05T 318C05T 319C05T K34 K108 K44 K45 K46 K47 214C 176CS01 177CS01 178CS01 179CS01
COPA310C25TEA 224 SGD
COPA310C25T COPA311C25T COPA312C25T COPA313C25T COPA316C05T COPA317C05T COPA318C05T COPA319C05T COPAK34 COPAK108 COPAK44 COPAK45 COPAK46 COPAK47 COPA214C COPA176CS01 COPA177CS01 COPA178CS01 COPA179CS01
Lab Consumables
Spreaders Inoculating Loops and Needles
A variety of high-quality lab consumables to meet laboratories everyday needs. We offer a wide range of transfer pipets, loops, spreaders, tubes, caps, and droppers meeting the highest quality standards. Since we produce them with our in-house technology, we can easily customise them upon customer’s request.
Streaking:
Loops and spreaders
Needles, spreaders, and inoculating loops fitting a multitude of different tasks
- Lubricants, oils, and electrostatic charges free
- Certified calibration
- Hexagonal loop shaft for easy handling
- Evans blue dye accuracy certification
Order Now
SPECIFICATIONS
Environmentally Preferable