SRK® Surfaces and Equipment Microbial Testing Devices
Supplier: Copan
902C 904C 905C 908C T2905C 922C 926C T2905CS01 T2906C T2906CS01 939CS01 940C 941C 929C 947C 961C 906C.CR 961CS01 952C
COPA902CEA 726 SGD
COPA902C COPA904C COPA905C COPA908C COPAT2905C COPA922C COPA926C COPAT2905CS01 COPAT2906C COPAT2906CS01 COPA939CS01 COPA940C COPA941C COPA929C COPA947C COPA961C COPA906C.CR COPA961CS01 COPA952C
SRK® Surfaces and Equipment Microbial Testing Devices
Swabs Swabs for Sampling
Our Swab Rinse Kits line includes comprehensive tailor-made collection and devices for the detection of environmental microbial contamination in a broad range of environments, such as food production, pharmaceutical sites, and hospital wards. Inspectors and Quality Assurance personnel can use SRK® products for routine surface monitoring within manufacturing areas, hospital surfaces, and equipment.
  • Optimal recovery rate: Choose the sampling probe that best suits your needs: regular FLOQSwabs® and CLASSIQSwabs™, plastic/viscose applicator, or sponge spatula
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable