384-well solid black polystyrene microplates, Corning®
Supplier: Corning
003654
CORN003654EA 0 SGD
CORN003654
384-well solid black polystyrene microplates, Corning®
Microplates HTS Microplates
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.