Erlenmeyer flask, PP, Corning®
Supplier: Corning
11410 11460
CORN11410EA 1510 SGD
CORN11410 CORN11460
Erlenmeyer flask, PP, Corning®
Flasks Erlenmeyer Flasks
These closed system erlenmeyer flasks are ideal to reduce contamination risks and enhance the productivity of your shaker culture operations when adding or removing medium and cells.
  • Chemically resistant, sterile
  • Heat sealable flexible tubing
  • Male luer lock disconnect
  • Cap colour: Orange
Flask is made from optically clear polycarbonate and is preassembled with an aseptic transfer cap.
Certifications: Meets USP, Class VI requirements for plastic containers and closures.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904