Aspirator bottles with tubing outlet, PYREX®
Supplier: Corning
1220-1L 1220-250 1220-2L 1220-2X 1220-3X 1220-4L 1220-5 1220-500
CORN1220-1LEA 113 SGD
CORN1220-1L CORN1220-250 CORN1220-2L CORN1220-2X CORN1220-3X CORN1220-4L CORN1220-5 CORN1220-500
Aspirator bottles with tubing outlet, PYREX®
Bottles Aspirators
PYREX®, borosilicate glass, without stoppers.
  • Excellent chemical resistance
  • High thermal resistance, autoclavable
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bottles with tubular sidearm outlet facilitates attachment of flexible tubing and it withstands repeated wet or dry autoclaving. It is designed for use as a delivery or storage container for solutions.
Order Now
SPECIFICATIONS
Environmentally Preferable