Cell culture bottles, with offset tooled neck, PYREX®
Supplier: Corning
1290-1L
CORN1290-1LEA 47.3 SGD
CORN1290-1L
Cell culture bottles, with offset tooled neck, PYREX®
Bottles Cell Culture Bottles
PYREX® borosilicate glass.
  • Offset neck is tool-finished for increased strength and uniform stopper fit
  • Useful for growing mass cultures
  • Withstand in repeated sterilization (wet or dry)
  • Stackable for storage
These culture bottles are designed so that when laid flat, it will hold approximately one-half of the stated capacity of solution.
Order Now
SPECIFICATIONS
Environmentally Preferable