Dropping bottles, with bulb and pipette stopper, PYREX®
Supplier: Corning
1340-125
CORN1340-125EA 825 SGD
CORN1340-125
Dropping bottles, with bulb and pipette stopper, PYREX®
Bottles Dropping Bottles
Narrow mouth droppings bottles, made of PYREX® borosilicate glass.
  • Reusable bottle
  • Pipette with suction bulb
  • Hollow glass stopper style
Recommended for use where preventing contamination by solubility or pH change is important.