Hypering bottle, graduated, PYREX®
Supplier: Corning
1350-2L
CORN1350-2LEA 330 SGD
CORN1350-2L
Hypering bottle, graduated, PYREX®
Bottles Cell Culture Bottles
Cylindrical, wide neck, borosilicate bottle has a tooled neck and graduations that can be read from top or bottom. They fit a no. 10 rubber stopper (not included).
  • Very good chemical resistance
  • Very good temperature resistance