Milk dilution bottles, with screw cap, PYREX®
Supplier: Corning
1367-160 1368-160 1372-160 1373-160
CORN1367-160EA 123 SGD
CORN1367-160 CORN1368-160 CORN1372-160 CORN1373-160
Milk dilution bottles, with screw cap, PYREX®
Bottles Dilution Bottles
Made of PYREX® borosilicate glass. Used for general storage of solutions and tissue culture work.
  • Wide mouth bottle offers easy measuring or filling
  • GPI 28-400 threaded black phenolic screw cap, supplied unattached, has a cemented-in rubber liner and is suitable for autoclaving
  • With a cut-line graduation mark at 99 ±1 ml
These bottles are designed from the requirements for milk dilution bottles given in the 'Standard Methods for the Examination of Dairy Products' published by the American Public Health Association.
Order Now
SPECIFICATIONS
Environmentally Preferable