Media bottles, graduated, PYREX®
Supplier: Corning
1395-150
CORN1395-150EA 211 SGD
CORN1395-150
Media bottles, graduated, PYREX®
Bottles Cell Culture Bottles
These PYREX® bottles are heavy-duty, reusable storage containers for media and other biological solutions.
  • Low-expansion borosilicate glass provides greater thermal strength
  • All parts withstand repeated autoclaving
Bottles have permanent white enamel graduations and a marking spot. Sizes 100 ml and above feature a glass bead that indicates full capacity line. Bottles are also ideal for dry heat sterilization (180 °C) when used with optional red PBT caps and red ETFE pouring rings.
Ordering information: Each bottle includes a linerless, one-piece,More Product Information