Roller bottle, PYREX®
Supplier: Corning
1400-285
CORN1400-285EA 322 SGD
CORN1400-285
Roller bottle, PYREX®
Bottles Roller Bottles
PYREX®, borosilicate glass.
  • Optical clarity and mechanical strength
  • Withstand wet or dry sterilisation
Delivery information: Supplied with 38 mm deep skirted GPI 38-430 threaded phenolic screw cap with rubber liner.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904