Weighing bottles, glass, PARR, PYREX®
Supplier: Corning
1684-2024
CORN1684-2024EA 508 SGD
CORN1684-2024
Weighing bottles, glass, PARR, PYREX®
Bottles Weighing Bottles
PYREX® borosilicate glass, cylindrical.
  • Excellent chemical resistance
  • Excellent temperature resistance
These Parr weighing bottles are designed for weighing small samples and have an external 24/12 standard taper joint style opening. The closure style cap fits over the external ground body.