Weighing bottle with external [ST] style opening, PYREX®
Supplier: Corning
1686-3060
CORN1686-3060EA 822 SGD
CORN1686-3060
Weighing bottle with external [ST] style opening, PYREX®
Bottles Weighing Bottles
These PYREX® standard weighing bottles have an external 29/12 Standard Taper joint style opening.
The closure style cap fits over the externally ground body.