Gas washing bottles, PYREX®
Supplier: Corning
1760-125 1760-250 1760-500
CORN1760-125EA 199 SGD
CORN1760-125 CORN1760-250 CORN1760-500
Gas washing bottles, PYREX®
Bottles Gas Washing Bottles
Made of PYREX®, borosilicate glass.
  • Humidify carbon dioxide-air mixtures for maintaining high humidity
  • Large hexagonal base for stability
  • Side arm tubulations are 8 mm Ext.Ø
Gas washing bottles are used for washing and drying gases. NS 29/42 stopper is used to seal the top to the base and it incorporates a plain tip tube as the distributor.
Order Now
SPECIFICATIONS
Environmentally Preferable