Nesbitt absorption bulb, PYREX®
Supplier: Corning
1900-165
CORN1900-165EA 461 SGD
CORN1900-165
Nesbitt absorption bulb, PYREX®
Bottles Gas Washing Bottles
Bulbs, made of PYREX® borosilicate glass are used for the absorption of CO₂ in carbon determination.
  • Weighs approximately 135 grams when filled
  • Reusable bulbs
  • A turn of the interchangeable stopper closes both inlet and outlet tubes (Ext.Ø 6 mm)