Condensers, Liebig, PYREX®
Supplier: Corning
2400-200 2400-300 2400-400
CORN2400-200EA 170 SGD
CORN2400-200 CORN2400-300 CORN2400-400
Condensers, Liebig, PYREX®
Condensers
Made of PYREX® borosilicate glass.
  • With outer joint at the top and inner standard taper joint with drip tip
  • Very good temperature resistance
  • Very good chemical resistance
This general purpose condenser can be used for distillation, refluxing and extraction operations. The condenser tubulations are approximately 10 mm Ext.Ø. Can be used for vacuum distillation.