Condensers, Allihn, PYREX®
Supplier: Corning
2480-200 2480-300 2480-400 2480-500 2480-600
CORN2480-200EA 234 SGD
CORN2480-200 CORN2480-300 CORN2480-400 CORN2480-500 CORN2480-600
Condensers, Allihn, PYREX®
Condensers
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These widely used 5 bulb condensers have a greater surface area than the corresponding Liebig type and are suitable for use in vacuum distillations. They have an outer standard taper joint at the top and an inner standard taper joint at the bottom with a drip tip. The tubulations have an Ø of approximately 10 mm.