Low volume microplates 96-well polystyrene, Corning® Stripwell™
Supplier: Corning
2481 2483 2485
CORN2481EA 2330 SGD
CORN2481 CORN2483 CORN2485
Low volume microplates 96-well polystyrene, Corning® Stripwell™
Microplates Immunology Microplates and Strips
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an ‘egg-crate’ style strip holder that allows everyone well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format
  • Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
  • Strips 1×8 are designed to fit only one way into the strip holder, eliminating the chance of misorientation
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk