Condensers, Graham, PYREX®
Supplier: Corning
2540-200 2540-300
CORN2540-200EA 283 SGD
CORN2540-200 CORN2540-300
Condensers, Graham, PYREX®
Condensers
Condensers made of PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These Graham condensers have a high surface area per unit with a standard taper outer joint at the top and a plain drip tip at the bottom. They are primarily used in distillation assemblies. Condenser tubulations are approximately 10 mm O.Ø.