Condensers, Graham, PYREX®
Supplier: Corning
2560-200 2560-300 2560-400 2560-500
CORN2560-200EA 269 SGD
CORN2560-200 CORN2560-300 CORN2560-400 CORN2560-500
Condensers, Graham, PYREX®
Condensers
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These Graham condensers have a high surface area per unit with a standard taper outer joint at the top and a standard taper inner joint at the bottom with a drip tip. They are primarily used in vacuum distillations. Condenser tubulations are approximately 10 mm ext.Ø.