Microscope slides, Corning®
Supplier: Corning
2947-75X25 2947-75X38 2947-75X50 2948-75X25 2949-75X25
CORN2947-75X25EA 591 SGD
CORN2947-75X25 CORN2947-75X38 CORN2947-75X50 CORN2948-75X25 CORN2949-75X25
Microscope slides, Corning®
Slides Microscope Slides
These slides are either frosted on one side, on both sides, or plain.
  • Clarity reinforced water-white glass
  • Frosted slides are sandblasted, easy to write on and highly legible
  • Each slide is inspected for chips and rough edge before being shipped