Corning® Disposable Anti-static Spatulas
Supplier: Corning
3003 3004 3005 3006 3007 3012 3013
CORN3003EA 492 SGD
CORN3003 CORN3004 CORN3005 CORN3006 CORN3007 CORN3012 CORN3013
Corning® Disposable Anti-static Spatulas
Spatulas
These spatulas are designed to save time and to provide contamination-free samples. They eliminate the recycling and resterilizing necessary with reusable spatulas.
  • Antistatic
  • Sterile
  • RNase-/DNase-free
  • Nonpyrogenic
Each spatula features two different tips to meet a variety of sampling requirements.
Ordering information: Available in five different configurations. Microspatulas available in two configurations.
Packaging: Individually packaged.