Corning® Storage Mats
Supplier: Corning
3080 3083 3341 3346
CORN3080EA 528 SGD
CORN3080 CORN3083 CORN3341 CORN3346
Corning® Storage Mats
Seals Microplate Seals
Multiple formats for specific and precise fit on 96-well and 384-well plates and blocks.
  • Non sterile
  • Not treated
Round well microplate storage mat: Fits Corning® 96 round well plates and blocks.
Manufactured from ethyl vinyl acetate (EVA), compatible with DMSO
Certified free from DNase and RNase
Can be applied manually or with applicator.
Square well storage block sealing mat: Fits Corning® 96 square 2 ml well storage blocks.
Manufactured from ethyl vinyl acetate (EVA), compatible with DMSO
Certified free from DNase and RNase
Can be applied manually orMore Product Information