Desiccators, with knob top, PYREX®
Supplier: Corning
3081-250 3081-150 3081-200
CORN3081-250EA 675 SGD
CORN3081-250 CORN3081-150 CORN3081-200
Desiccators, with knob top, PYREX®
Desiccators Desiccators, Glass
These PYREX® borosilicate glass desiccators are designed for general analytical work. Material is Borosilicate Glass Code 7740.
  • Sturdy knobs make the lids easy to handle
Ordering information: Plate is not included. For a compatible plate, please contact your representative for more information.
Caution: Warning: Do not heat or subject to pressure. Not suitable for vacuum applications.
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing