Dishes, crystallising, Borosilicate glass, PYREX®
Supplier: Corning
3140-100 3140-125 3140-150 3140-170 3140-190 3140-70 3140-80 3140-90
CORN3140-100EA 361 SGD
CORN3140-100 CORN3140-125 CORN3140-150 CORN3140-170 CORN3140-190 CORN3140-70 CORN3140-80 CORN3140-90
Dishes, crystallising, Borosilicate glass, PYREX®
Trays/Dishes/Pots Crystallising Dishes
PYREX® borosilicate glass, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.
Order Now
SPECIFICATIONS
Environmentally Preferable