Heavy wall drying trays, PYREX®
Supplier: Corning
3170-12
CORN3170-12EA 304 SGD
CORN3170-12
Heavy wall drying trays, PYREX®
Trays/Dishes/Pots Drying Trays
These heavy wall dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.
  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean
They can be used with gel electrophoresis or paper chromatography.
Caution: For oven use only. Do not use over direct heat.