Gas washing bottle, with side inlet, coarse fritted disc, PYREX®
Supplier: Corning
31750-350C
CORN31750-350CEA 541 SGD
CORN31750-350C
Gas washing bottle, with side inlet, coarse fritted disc, PYREX®
Bottles Gas Washing Bottles
PYREX® borosilicate glass.
  • Humidify carbon dioxide-air mixtures for maintaining high humidity
  • Large diameter fritted disc permits the handling of large volumes of gas
  • Fritted disc provides greater efficiency and more uniform dispersion of gas bubbles for complete absorption
These reusable gas washing bottles are used for washing and drying gases. The side inlet tube on this bottle has a rod sealed to the bottle near the top for extra strength. NS 24/40 stopper is used to seal the outlet tube to the base. Tubulation Outer dimension is 10 mm. CoarseMore Product Information