IVF culture dish, Costar®
Supplier: Corning
3260
CORN3260EA 1260 SGD
CORN3260
IVF culture dish, Costar®
Culture Dishes Cell Culture Dishes
Optically clear culture dish with 20 mm centre well. Inner well holds 3 ml of medium while the outer well holds 10 ml.
  • 45° slope funnels oocyte/embryo into centre of well
  • Stand-off rim on base protects optical surface from scratching
  • Treated for optimal cell attachment
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
Product is strictly not for human use. The center well dish is a Class I medical device and should never be used in applications which require the product to be certified as a Class II or Class III medical device.
Center wellMore Product Information