Cell culture chambers, Corning® CellSTACK®
Supplier: Corning
3268 3269 3270 3271 3272 3319 3310 3320 3321 3330 3311 3312 3303 3313
CORN3268EA 1060 SGD
CORN3268 CORN3269 CORN3270 CORN3271 CORN3272 CORN3319 CORN3310 CORN3320 CORN3321 CORN3330 CORN3311 CORN3312 CORN3303 CORN3313
Cell culture chambers, Corning® CellSTACK®
Bioreactors
PS, sterile. The Corning® CellSTACK® culture chambers are available in five sizes and with three different surface treatments.
  • Two 26 mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system
  • Standard 33 mm threaded caps have 0,2 μm pore non-wettable membranes sealed directly to the caps to allow gas exchange while minimising the risk of contamination
  • Optional 33 mm threaded caps are available with integrally sealed USP Class VI
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk