24 clear not treated reservoir, sterile, HTS Transwell®
Supplier: Corning
3395
CORN3395EA 326 SGD
CORN3395
24 clear not treated reservoir, sterile, HTS Transwell®
Microplates HTS Microplates
For use as reservoir tray with HTS Transwell permeable supports.
  • Not treated for cell attachment
  • Same footprint as standard 24 well plate but with single rectangular well