Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
3401 3402 3412 3413 3414 3415 3420 3421 3422 3428
CORN3401EA 480 SGD
CORN3401 CORN3402 CORN3412 CORN3413 CORN3414 CORN3415 CORN3420 CORN3421 CORN3422 CORN3428
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Microplates Insert Plates and Inserts
75 mm Transwell® with 0,4 µm pore polycarbonate membrane insert, sterile.
  • 10 µm thick translucent polycarbonate membrane
  • Treated for optimal cell attachment
  • Each insert is packaged in a 100 mm dish, 12 inserts per case
  • Membrane must be stained for cell visibility
Produces a cell culture environment that closely resembles an in vivo state. An essential tool for the study of both anchorage-dependent and independent cell lines.